Order Entry
Czech Republic
ContactUsLinkComponent
4-Methoxy-3-(trifluoromethyl)benzyl alcohol 97%
Irritant
4-Methoxy-3-(trifluoromethyl)benzyl alcohol 97%
Catalog # APOSPC0410-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Methoxy-3-(trifluoromethyl)benzyl alcohol 97%
Catalog # APOSPC0410-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :206.162 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:261951-88-6
Formula:C₉H₉F₃O₂
MDL Number:MFCD01631577