Order Entry
Czech Republic
ContactUsLinkComponent
2-(Difluoromethylthio)benzyl alcohol
2-(Difluoromethylthio)benzyl alcohol
Catalog # APOSPC0279-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(Difluoromethylthio)benzyl alcohol
Catalog # APOSPC0279-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :190.21 g/mol
Storage Temperature:Ambient
CAS Number:261944-18-7
Formula:C₈H₈F₂OS
MDL Number:MFCD01631520