
1-[3-(Difluoromethoxy)phenyl]imidazolidin-2-one
Catalog # APOSPC01478-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:228.195 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1378261-25-6
Formula:C₁₀H₁₀F₂N₂O₂
MDL Number:MFCD20037413