
6-Nitrochroman-4-one 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:68043-53-8
:
Formula:C₉H₇NO₄
MDL Number:MFCD04038185
:193.156 g/mol
Melting Pt:116…119 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together