Order Entry
Czech Republic
ContactUsLinkComponent
2-Chloro-1-(4-pyridinyl)ethanone hydrochloride 95%
2-Chloro-1-(4-pyridinyl)ethanone hydrochloride 95%
Catalog # APOSOR965660-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-1-(4-pyridinyl)ethanone hydrochloride 95%
Catalog # APOSOR965660-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :192.043 g/mol
Storage Temperature:Ambient
CAS Number:25260-36-0
Formula:C₇H₆ClNO·ClH
MDL Number:MFCD08692051