Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Expresní objednávka
Stav objednávky
Váš účet
Košík
Czech Republic
Expresní objednávka
Stav objednávky
Váš účet
Košík
ContactUsLinkComponent
Home
Methyl 2-(1-methylpiperidin-4-yl)acetate
Methyl 2-(1-methylpiperidin-4-yl)acetate 98%
Methyl 2-(1-methylpiperidin-4-yl)acetate 98%
APOSOR964903-1G
:
Apollo Scientific
:
Methyl 2-(1-methylpiperidin-4-yl)acetate 98%
APOSOR964903-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
171.237 g/mol
:
Ambient
:
95533-25-8
:
C₉H₁₇NO₂
:
MFCD12756507