5-Methyl-2-phenyl-3-furoyl chloride 97%
Catalog # APOSOR9643-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:220.652 g/mol
Boiling Pt:120…125 °C (0.5 mm Hg)
Storage Temperature:Ambient
CAS Number:183210-32-4
Formula:C₁₂H₉ClO₂
MDL Number:MFCD08435904