Order Entry
Czech Republic
ContactUsLinkComponent
2-Methoxy-4-(2-nitroethyl)phenol 95%
2-Methoxy-4-(2-nitroethyl)phenol 95%
Catalog # APOSOR963988-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Methoxy-4-(2-nitroethyl)phenol 95%
Catalog # APOSOR963988-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :197.188 g/mol
Storage Temperature:Ambient
CAS Number:528594-30-1
Formula:C₉H₁₁NO₄
MDL Number:MFCD03410699