2-Chloro-5-(1-methyl-1H-pyrazol-4-yl)pyrimidine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1231943-08-0
:
Formula:C₈H₇ClN₄
MDL Number:MFCD17926235
:194.621 g/mol
Storage Temperature:Ambient
Frequently Bought Together