
2-(4-Bromophenyl)ethanamine, N-BOC protected 97%
Catalog # APOSOR962698-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:300.192 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:120157-97-3
Formula:C₁₃H₁₈BrNO₂
MDL Number:MFCD08443684