Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Methoxy-4,6-dimethylpyrimidine
2-Methoxy-4,6-dimethylpyrimidine 95%
2-Methoxy-4,6-dimethylpyrimidine 95%
APOSOR961880-1G
:
Apollo Scientific
:
2-Methoxy-4,6-dimethylpyrimidine 95%
APOSOR961880-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
138.167 g/mol
:
Ambient
:
14001-61-7
:
C₇H₁₀N₂O
:
MFCD02765642