1-Methyl-1H-benzimidazol-5-ol 98%
Catalog # APOSOR961057-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:148.162 g/mol
Storage Temperature:Ambient
CAS Number:50591-22-5
Formula:C₈H₈N₂O
MDL Number:MFCD00456136