Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fmoc-cys(npys)-oh
Fmoc-cys(npys)-oh
Fmoc-cys(npys)-oh
Fmoc-cys(npys)-oh
APOSOR960649-1G
:
Apollo Scientific
:
Fmoc-cys(npys)-oh
APOSOR960649-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
497.543 g/mol
:
GHS07
:
Ambient
:
159700-51-3
:
C₂₃H₁₉N₃O₆S₂
:
MFCD00065638