Order Entry
Czech Republic
ContactUsLinkComponent
1-Isoxazol-5-yl-ethylamine hydrochloride 95%
1-Isoxazol-5-yl-ethylamine hydrochloride 95%
Catalog # APOSOR960585-100MG
Supplier:  Apollo Scientific
CAS Number:  
1-Isoxazol-5-yl-ethylamine hydrochloride 95%
Catalog # APOSOR960585-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :148.591 g/mol
Storage Temperature:Ambient
CAS Number:1187930-08-0
Formula:C₅H₈N₂O·ClH
MDL Number:MFCD06738810