Order Entry
Czech Republic
ContactUsLinkComponent
2-Ethoxy-3-methylpyridine-5-carboxylic acid 95%
2-Ethoxy-3-methylpyridine-5-carboxylic acid 95%
Catalog # APOSOR960260-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Ethoxy-3-methylpyridine-5-carboxylic acid 95%
Catalog # APOSOR960260-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :181.189 g/mol
Storage Temperature:Ambient
CAS Number:1355216-85-1
Formula:C₉H₁₁NO₃
MDL Number:MFCD21087314