Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-(3-Formylpyridin-2-yl)acetamide
N-(3-Formylpyridin-2-yl)acetamide 97%
N-(3-Formylpyridin-2-yl)acetamide 97%
APOSOR959719-1G
:
Apollo Scientific
:
N-(3-Formylpyridin-2-yl)acetamide 97%
APOSOR959719-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
164.161 g/mol
:
Ambient
:
54856-85-8
:
C₈H₈N₂O₂
:
MFCD17170608