Order Entry
Czech Republic
ContactUsLinkComponent
cis-1,2-Cyclohexanedicarboximide 95%
cis-1,2-Cyclohexanedicarboximide 95%
Catalog # APOSOR959262-100G
Supplier:  Apollo Scientific
CAS Number:  
cis-1,2-Cyclohexanedicarboximide 95%
Catalog # APOSOR959262-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :153.178 g/mol
Storage Temperature:Ambient
CAS Number:7506-66-3
Formula:C₈H₁₁NO₂
MDL Number:MFCD00067706