N1,N2-Dimethylcyclohexane-1,2-diamine hydrochloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1956325-54-4
:
Formula:C₈H₁₈N₂·ClH
MDL Number:MFCD21603997
:178.703 g/mol
Storage Temperature:Ambient
Frequently Bought Together