Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(Dimethylamino)-3-nitrophenol
4-(Dimethylamino)-3-nitrophenol 95%
4-(Dimethylamino)-3-nitrophenol 95%
APOSOR957814-1G
:
Apollo Scientific
:
4-(Dimethylamino)-3-nitrophenol 95%
APOSOR957814-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
182.177 g/mol
:
Ambient
:
14703-89-0
:
C₈H₁₀N₂O₃
:
MFCD20926279