Order Entry
Czech Republic
ContactUsLinkComponent
4-Ethoxy-3-nitrophenol 95%
4-Ethoxy-3-nitrophenol 95%
Catalog # APOSOR957779-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Ethoxy-3-nitrophenol 95%
Catalog # APOSOR957779-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :183.161 g/mol
Storage Temperature:Ambient
CAS Number:1394969-99-3
Formula:C₈H₉NO₄
MDL Number:MFCD23111049