Ethyl 3-hydroxy-4-methoxybenzoate 95%
Catalog # APOSOR957711-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:196.2 g/mol
Storage Temperature:Ambient
CAS Number:148527-38-2
Formula:C₁₀H₁₂O₄
MDL Number:MFCD06204303