Order Entry
Czech Republic
ContactUsLinkComponent
 
N,N-Diethyl-3-hydroxybenzamide 95%
Catalog # APOSOR957705-1G
Supplier:  Apollo Scientific
CAS Number:  
N,N-Diethyl-3-hydroxybenzamide 95%
Catalog # APOSOR957705-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :193.242 g/mol
Storage Temperature:Ambient
CAS Number:15789-04-5
Formula:C₁₁H₁₅NO₂
MDL Number:MFCD11130615