Order Entry
Czech Republic
ContactUsLinkComponent
4,5-Dichloro-2-methoxyphenol 98%
4,5-Dichloro-2-methoxyphenol 98%
Catalog # APOSOR957647-1G
Supplier:  Apollo Scientific
CAS Number:  
4,5-Dichloro-2-methoxyphenol 98%
Catalog # APOSOR957647-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :193.027 g/mol
Storage Temperature:Ambient
CAS Number:2460-49-3
Formula:C₇H₆Cl₂O₂
MDL Number:MFCD01707578