Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl 2-(4-hydroxyphenoxy)acetate 95%
Ethyl 2-(4-hydroxyphenoxy)acetate 95%
Catalog # APOSOR957644-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 2-(4-hydroxyphenoxy)acetate 95%
Catalog # APOSOR957644-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :196.2 g/mol
Storage Temperature:Ambient
CAS Number:20872-28-0
Formula:C₁₀H₁₂O₄
MDL Number:MFCD12025129