Order Entry
Czech Republic
ContactUsLinkComponent
1-(4-Chloro-3-hydroxyphenyl)ethanone 98%
1-(4-Chloro-3-hydroxyphenyl)ethanone 98%
Catalog # APOSOR957632-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(4-Chloro-3-hydroxyphenyl)ethanone 98%
Catalog # APOSOR957632-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :170.593 g/mol
Storage Temperature:Ambient
CAS Number:61124-56-9
Formula:C₈H₇ClO₂
MDL Number:MFCD00034670