Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-[(Cyclopentylamino)methyl]phenol
2-[(Cyclopentylamino)methyl]phenol 95%
2-[(Cyclopentylamino)methyl]phenol 95%
2-[(Cyclopentylamino)methyl]phenol 95%
APOSOR957620-1G
:
Apollo Scientific
:
2-[(Cyclopentylamino)methyl]phenol 95%
APOSOR957620-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
191.269 g/mol
:
Ambient
:
827328-86-9
:
C₁₂H₁₇NO
:
MFCD11139824