Order Entry
Czech Republic
ContactUsLinkComponent
2-(4-Hydroxy-2,6-dimethylphenoxy)acetonitrile 95%
2-(4-Hydroxy-2,6-dimethylphenoxy)acetonitrile 95%
Catalog # APOSOR957617-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(4-Hydroxy-2,6-dimethylphenoxy)acetonitrile 95%
Catalog # APOSOR957617-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :177.2 g/mol
Storage Temperature:Ambient
CAS Number:1394917-42-0
Formula:C₁₀H₁₁NO₂
MDL Number:MFCD23111033