Order Entry
Czech Republic
ContactUsLinkComponent
4-Methyl-1-naphthol 95%
4-Methyl-1-naphthol 95%
Catalog # APOSOR957594-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Methyl-1-naphthol 95%
Catalog # APOSOR957594-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :158.196 g/mol
Storage Temperature:Ambient
CAS Number:10240-08-1
Formula:C₁₁H₁₀O
MDL Number:MFCD00060344