Order Entry
Czech Republic
ContactUsLinkComponent
4-(Cyclopent-1-en-1-yl)phenol 95%
4-(Cyclopent-1-en-1-yl)phenol 95%
Catalog # APOSOR957590-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(Cyclopent-1-en-1-yl)phenol 95%
Catalog # APOSOR957590-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :160.212 g/mol
Storage Temperature:Ambient
Formula:C₁₁H₁₂O
MDL Number:MFCD23111005