Order Entry
Czech Republic
ContactUsLinkComponent
4-Chloro-3-propoxyphenol 95%
4-Chloro-3-propoxyphenol 95%
Catalog # APOSOR957573-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-3-propoxyphenol 95%
Catalog # APOSOR957573-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :186.635 g/mol
Storage Temperature:Ambient
CAS Number:1378655-26-5
Formula:C₉H₁₁ClO₂
MDL Number:MFCD19440097