Order Entry
Czech Republic
ContactUsLinkComponent
2'-Chloro-5'-hydroxyacetophenone 95%
2'-Chloro-5'-hydroxyacetophenone 95%
Catalog # APOSOR957566-1G
Supplier:  Apollo Scientific
CAS Number:  
2'-Chloro-5'-hydroxyacetophenone 95%
Catalog # APOSOR957566-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :170.593 g/mol
Storage Temperature:Ambient
CAS Number:58020-38-5
Formula:C₈H₇ClO₂
MDL Number:MFCD18393141