Order Entry
Czech Republic
ContactUsLinkComponent
4-Methoxy-2,5-dimethylphenol 95%
4-Methoxy-2,5-dimethylphenol 95%
Catalog # APOSOR957564-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Methoxy-2,5-dimethylphenol 95%
Catalog # APOSOR957564-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :152.19 g/mol
Storage Temperature:Ambient
CAS Number:4962-28-1
Formula:C₉H₁₂O₂
MDL Number:MFCD14537875