Order Entry
Czech Republic
ContactUsLinkComponent
 
2,6-Dichloro-3-methylphenol 95%
Catalog # APOSOR957526-1G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Dichloro-3-methylphenol 95%
Catalog # APOSOR957526-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :177.028 g/mol
Storage Temperature:Ambient
CAS Number:13481-70-4
Formula:C₇H₆Cl₂O
MDL Number:MFCD08061909