Order Entry
Czech Republic
ContactUsLinkComponent
1-(Oxan-2-yl)-1H-pyrazol-4-ol 95%
1-(Oxan-2-yl)-1H-pyrazol-4-ol 95%
Catalog # APOSOR957523-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(Oxan-2-yl)-1H-pyrazol-4-ol 95%
Catalog # APOSOR957523-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :168.193 g/mol
Storage Temperature:Ambient
Formula:C₈H₁₂N₂O₂
MDL Number:MFCD23111248