4-[(Dimethylamino)methyl]phenol 95%
Catalog # APOSOR957515-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:151.206 g/mol
Storage Temperature:Ambient
CAS Number:103-87-7
Formula:C₉H₁₃NO
MDL Number:MFCD00053408