Order Entry
Czech Republic
ContactUsLinkComponent
2-(3-Hydroxyphenoxy)acetonitrile 95%
2-(3-Hydroxyphenoxy)acetonitrile 95%
Catalog # APOSOR957488-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(3-Hydroxyphenoxy)acetonitrile 95%
Catalog # APOSOR957488-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :149.147 g/mol
Storage Temperature:Ambient
CAS Number:107223-69-8
Formula:C₈H₇NO₂
MDL Number:MFCD11181964