Order Entry
Czech Republic
ContactUsLinkComponent
3-(2-Methoxyethoxy)phenol 98%
3-(2-Methoxyethoxy)phenol 98%
Catalog # APOSOR957485-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(2-Methoxyethoxy)phenol 98%
Catalog # APOSOR957485-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :168.19 g/mol
Storage Temperature:Ambient
CAS Number:245070-91-1
Formula:C₉H₁₂O₃
MDL Number:MFCD11103459