Order Entry
Czech Republic
ContactUsLinkComponent
2-(Dimethylamino)pyrimidin-5-ol 95%
2-(Dimethylamino)pyrimidin-5-ol 95%
Catalog # APOSOR957481-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(Dimethylamino)pyrimidin-5-ol 95%
Catalog # APOSOR957481-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :139.155 g/mol
Storage Temperature:Ambient
CAS Number:72491-71-5
Formula:C₆H₉N₃O
MDL Number:MFCD10696945