Order Entry
Czech Republic
ContactUsLinkComponent
6-(Dimethylamino)pyridin-3-ol 95%
6-(Dimethylamino)pyridin-3-ol 95%
Catalog # APOSOR957480-1G
Supplier:  Apollo Scientific
CAS Number:  
6-(Dimethylamino)pyridin-3-ol 95%
Catalog # APOSOR957480-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :138.167 g/mol
Storage Temperature:Ambient
CAS Number:330473-71-7
Formula:C₇H₁₀N₂O
MDL Number:MFCD16998105