Order Entry
Czech Republic
ContactUsLinkComponent
4-Ethoxy-3-methylphenol 95%
4-Ethoxy-3-methylphenol 95%
Catalog # APOSOR957457-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Ethoxy-3-methylphenol 95%
Catalog # APOSOR957457-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :152.19 g/mol
Storage Temperature:Ambient
CAS Number:1216257-46-3
Formula:C₉H₁₂O₂
MDL Number:MFCD14535356