1,3-dimethyl-1H-pyrazol-4-ol 95%
Catalog # APOSOR957431-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:112.13 g/mol
Storage Temperature:Ambient
CAS Number:85985-66-6
Formula:C₅H₈N₂O
MDL Number:MFCD23110698