Order Entry
Czech Republic
ContactUsLinkComponent
1-Butyl-1H-pyrazol-4-ol 95%
1-Butyl-1H-pyrazol-4-ol 95%
Catalog # APOSOR957421-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Butyl-1H-pyrazol-4-ol 95%
Catalog # APOSOR957421-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :140.183 g/mol
Storage Temperature:Ambient
Formula:C₇H₁₂N₂O
MDL Number:MFCD23110964