0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Ethyl (4-chloro-1H-pyrazol-1-yl)acetate
Ethyl (4-chloro-1H-pyrazol-1-yl)acetate 95%
Ethyl (4-chloro-1H-pyrazol-1-yl)acetate 95%
APOSOR957054-1G
:
Apollo Scientific
:
Ethyl (4-chloro-1H-pyrazol-1-yl)acetate 95%
APOSOR957054-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
188.612 g/mol
:
Ambient
:
C₇H₉ClN₂O₂
:
MFCD02874251