5-Bromo-N-cyclopropyl-2-methoxybenzenesulphonamide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1000339-35-4
:914302-35-5
Formula:C₁₀H₁₂BrNO₃S
MDL Number:MFCD08235093
:306.176 g/mol
Melting Pt:134…136 °C
Storage Temperature:Refrigerator
Frequently Bought Together