Order Entry
Czech Republic
ContactUsLinkComponent
1-Chloro-4-(2-methoxyethoxy)benzene 95%
1-Chloro-4-(2-methoxyethoxy)benzene 95%
Catalog # APOSOR956993-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Chloro-4-(2-methoxyethoxy)benzene 95%
Catalog # APOSOR956993-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :186.635 g/mol
Storage Temperature:Ambient
CAS Number:60893-72-3
Formula:C₉H₁₁ClO₂
MDL Number:MFCD20390459