Order Entry
Czech Republic
ContactUsLinkComponent
5-Chloro-2-dimethylaminopyrimidine 95%
5-Chloro-2-dimethylaminopyrimidine 95%
Catalog # APOSOR956989-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Chloro-2-dimethylaminopyrimidine 95%
Catalog # APOSOR956989-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :157.601 g/mol
Storage Temperature:Ambient
CAS Number:81568-09-4
Formula:C₆H₈ClN₃
MDL Number:MFCD16657234