4-Chloro-1-isobutyl-1H-pyrazole 95%
Catalog # APOSOR956958-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:158.629 g/mol
Storage Temperature:Ambient
CAS Number:1204901-78-9
Formula:C₇H₁₁ClN₂
MDL Number:MFCD15976271