Order Entry
Czech Republic
ContactUsLinkComponent
5-Chloro-2-methoxy-3-methylpyridine 95%
5-Chloro-2-methoxy-3-methylpyridine 95%
Catalog # APOSOR956947-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Chloro-2-methoxy-3-methylpyridine 95%
Catalog # APOSOR956947-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :157.598 g/mol
Storage Temperature:Ambient
CAS Number:1261488-23-6
Formula:C₇H₈ClNO
MDL Number:MFCD18415742