Order Entry
Czech Republic
ContactUsLinkComponent
3-Chlorocyclohexan-1-one 95%
3-Chlorocyclohexan-1-one 95%
Catalog # APOSOR956918-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Chlorocyclohexan-1-one 95%
Catalog # APOSOR956918-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :132.588 g/mol
Storage Temperature:Ambient
Formula:C₆H₉ClO
MDL Number:MFCD20621602