5-Nitro-1-propyl-1H-pyrazole 95%
Catalog # APOSOR956255-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:155.155 g/mol
Storage Temperature:Ambient
CAS Number:1170522-30-1
Formula:C₆H₉N₃O₂
MDL Number:MFCD09468158